Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2-methyl-6-nitroaniline, 97%

Click to view available options
Quantity:
1g
5g
25g
77811-44-0
C7H7BrN2O2
231.049
MFCD00052919
ZXFVKFUXKFPUQJ-UHFFFAOYSA-N
4-bromo-2-methyl-6-nitro-phenylamine, 4-bromo-2-methyl-6-nitrobenzenamine, 2-amino-5-bromo-3-nitrotoluene, 4-bromo-6-nitro-o-toluidine, benzenamine, 4-bromo-2-methyl-6-nitro, 4-bromo-2-methyl-6-nitrophenylamine, 4-bromo-2-methyl-6-nitro-aniline, pubchem3813, acmc-1bg8u, ksc496e7t
522679
4-bromo-2-methyl-6-nitroaniline
CC1=CC(=CC(=C1N)[N+](=O)[O-])Br
77811-44-0
C7H7BrN2O2
231.049
MFCD00052919
ZXFVKFUXKFPUQJ-UHFFFAOYSA-N
4-bromo-2-methyl-6-nitro-phenylamine, 4-bromo-2-methyl-6-nitrobenzenamine, 2-amino-5-bromo-3-nitrotoluene, 4-bromo-6-nitro-o-toluidine, benzenamine, 4-bromo-2-methyl-6-nitro, 4-bromo-2-methyl-6-nitrophenylamine, 4-bromo-2-methyl-6-nitro-aniline, pubchem3813, acmc-1bg8u, ksc496e7t
522679
4-bromo-2-methyl-6-nitroaniline
CC1=CC(=CC(=C1N)[N+](=O)[O-])Br
Applications
It is used in the design and synthesis of CK2 inhibitors and in the docking studies of novel telmisartan-glitazone hybrid analogs for the treatment of metabolic syndrome.

Solubility
Slightly soluble in water and soluble in hot methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Quantity 1g
Melting Point 143°C to 147°C
Beilstein 3530524
Solubility Information Slightly soluble in water and soluble in hot methanol.
Formula Weight 231.06
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methyl-6-nitroaniline

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures