Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2-methylaniline, 98%

Click to view available options
Quantity:
25g
100g
583-75-5
C7H8BrN
186.052
MFCD00007825
PCHYYOCUCGCSBU-UHFFFAOYSA-N
2-amino-5-bromotoluene,4-bromo-o-toluidine,2-methyl-4-bromoaniline,benzenamine, 4-bromo-2-methyl,o-toluidine, 4-bromo,4-bromo-2-methyl-phenylamine,4-bromo-2-methylbenzenamine,5-bromo-2-aminotoluene,1-amino-4-bromo-2-methylbenzene,4-bromo-2-methylbenenamine
11423
4-bromo-2-methylaniline
CC1=C(C=CC(=C1)Br)N
583-75-5
C7H8BrN
186.052
MFCD00007825
PCHYYOCUCGCSBU-UHFFFAOYSA-N
2-amino-5-bromotoluene,4-bromo-o-toluidine,2-methyl-4-bromoaniline,benzenamine, 4-bromo-2-methyl,o-toluidine, 4-bromo,4-bromo-2-methyl-phenylamine,4-bromo-2-methylbenzenamine,5-bromo-2-aminotoluene,1-amino-4-bromo-2-methylbenzene,4-bromo-2-methylbenenamine
11423
4-bromo-2-methylaniline
CC1=C(C=CC(=C1)Br)N
Odor Characteristic
Flash Point >110°C (230°F)
Quantity 25g
Melting Point 56°C to 60°C
Boiling Point 240°C
UN Number UN2811
Beilstein 636521
Formula Weight 186.06
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-methylaniline

GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c

H315-H319-H335-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 209-519-9

RTECSNumber : XU4250000

TSCA : Yes

Recommended Storage : Ambient temperatures