Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-methylbenzonitrile, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
67832-11-5
C8H6BrN
196.047
MFCD00041461
LPEBMDFRIKYFCF-UHFFFAOYSA-N
benzonitrile, 4-bromo-2-methyl,5-bromo-2-cyanotoluene,4-bromo-2-methyl-benzonitrile,2-methyl-4-bromo benzonitrile,2-methyl-4-bromobenzonitrile,4-bromo-2-methylbenzene carbonitrile,4-bromo-2-methylbenzenecarbonitrile,pubchem4721,4-bromo-methylbenzonitrile,4bromo-2-methylbenzonitrile
144267
4-bromo-2-methylbenzonitrile
CC1=C(C=CC(=C1)Br)C#N
67832-11-5
C8H6BrN
196.047
MFCD00041461
LPEBMDFRIKYFCF-UHFFFAOYSA-N
benzonitrile, 4-bromo-2-methyl,5-bromo-2-cyanotoluene,4-bromo-2-methyl-benzonitrile,2-methyl-4-bromo benzonitrile,2-methyl-4-bromobenzonitrile,4-bromo-2-methylbenzene carbonitrile,4-bromo-2-methylbenzenecarbonitrile,pubchem4721,4-bromo-methylbenzonitrile,4bromo-2-methylbenzonitrile
144267
4-bromo-2-methylbenzonitrile
CC1=C(C=CC(=C1)Br)C#N
Quantity 5g
Melting Point 63°C to 68°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color White to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 196.04
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 4-Bromo-2-methylbenzonitrile, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning