Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2-nitroanisole, 97%

Click to view available options
Quantity:
5g
25g
100g
33696-00-3
C7H6BrNO3
232.033
MFCD00055529
ORBHQHXVVMZIDP-UHFFFAOYSA-N
4-bromo-2-nitroanisole,2-methoxy-5-bromonitrobenzene,4-brom-2-nitroanisole,4-bromo-2-nitro anisole,5-bromo-2-methoxynitrobenzene,3-nitro-4-methoxy bromobenzene,1-methoxy-2-nitro-4-bromobenzene,benzene, 4-bromo-1-methoxy-2-nitro,pubchem5341,4-brom-2-nitroanisol
118533
4-bromo-1-methoxy-2-nitrobenzene
COC1=C(C=C(C=C1)Br)[N+](=O)[O-]
33696-00-3
C7H6BrNO3
232.033
MFCD00055529
ORBHQHXVVMZIDP-UHFFFAOYSA-N
4-bromo-2-nitroanisole,2-methoxy-5-bromonitrobenzene,4-brom-2-nitroanisole,4-bromo-2-nitro anisole,5-bromo-2-methoxynitrobenzene,3-nitro-4-methoxy bromobenzene,1-methoxy-2-nitro-4-bromobenzene,benzene, 4-bromo-1-methoxy-2-nitro,pubchem5341,4-brom-2-nitroanisol
118533
4-bromo-1-methoxy-2-nitrobenzene
COC1=C(C=C(C=C1)Br)[N+](=O)[O-]
Quantity 5g
Melting Point 85°C to 100°C
Beilstein 2259903
Formula Weight 232.03
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-nitroanisole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C