Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-nitrophenol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
7693-52-9
C6H4BrNO3
218.01
MFCD00082540
CUTFAPGINUFNQM-UHFFFAOYSA-N
2-nitro-4-bromophenol,phenol, 4-bromo-2-nitro,4-bromo-2-nitro-phenol,4-bromo-1-hydroxy-2-nitrobenzene,phenol, 2-nitro-5-bromo,pubchem4099,4-brom-2-nitro-phenol,acmc-209p6y,4-06-00-01363 beilstein handbook reference,ksc494e2j
24364
4-bromo-2-nitrophenol
C1=CC(=C(C=C1Br)[N+](=O)[O-])O
7693-52-9
C6H4BrNO3
218.01
MFCD00082540
CUTFAPGINUFNQM-UHFFFAOYSA-N
2-nitro-4-bromophenol,phenol, 4-bromo-2-nitro,4-bromo-2-nitro-phenol,4-bromo-1-hydroxy-2-nitrobenzene,phenol, 2-nitro-5-bromo,pubchem4099,4-brom-2-nitro-phenol,acmc-209p6y,4-06-00-01363 beilstein handbook reference,ksc494e2j
24364
4-bromo-2-nitrophenol
C1=CC(=C(C=C1Br)[N+](=O)[O-])O
Quantity 25g
Color Yellow
Packaging Glass bottle
Melting Point 87.0°C to 94.0°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula BrC6H3(NO2)OH
Beilstein 06, IV, 1363
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,acetone,toluene
Formula Weight 218.01
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-nitrophenol, 98%
Show More Show Less

GHS H Statement
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
W

GHS Signal Word: Warning

EINECSNumber : 231-707-4