Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2-nitrotoluene, 99%

Click to view available options
Quantity:
5g
25g
100g
60956-26-5
C7H6BrNO2
216.034
MFCD00041243
KZNXALJXBRSMFL-UHFFFAOYSA-N
4-bromo-2-nitrotoluene, 2-nitro-4-bromotoluene, 4-bromo-6-nitrotoluene, bromo 4--2-nitrotoluene, 5-bromo-2-methylnitrobenzene, benzene, 4-bromo-1-methyl-2-nitro, 4-bromo-1-methyl-2-nitro-benzene, 4-brom-2-nitrotoluol, pubchem10403, maybridge1_002072
123546
4-bromo-1-methyl-2-nitrobenzene
CC1=C(C=C(C=C1)Br)[N+](=O)[O-]
60956-26-5
C7H6BrNO2
216.034
MFCD00041243
KZNXALJXBRSMFL-UHFFFAOYSA-N
4-bromo-2-nitrotoluene, 2-nitro-4-bromotoluene, 4-bromo-6-nitrotoluene, bromo 4--2-nitrotoluene, 5-bromo-2-methylnitrobenzene, benzene, 4-bromo-1-methyl-2-nitro, 4-bromo-1-methyl-2-nitro-benzene, 4-brom-2-nitrotoluol, pubchem10403, maybridge1_002072
123546
4-bromo-1-methyl-2-nitrobenzene
CC1=C(C=C(C=C1)Br)[N+](=O)[O-]
Flash Point >110°C (230°F)
Boiling Point 130°C (12mmHg)
Melting Point 45°C to 48°C
Quantity 5g
Beilstein 2328190
Formula Weight 216.04
Percent Purity 99%
Chemical Name or Material 4-Bromo-2-nitrotoluene

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 262-536-3

TSCA : No

Recommended Storage : Ambient temperatures