Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 4-Bromo-2-(trifluoromethoxy)phenol, 98%
Click to view available options
Quantity:
1g
5g
Description
Applications
4-Bromo-2-(trifluoromethoxy)phenol are the basic building blocks numerous pharmaceutical drugs.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
4-Bromo-2-(trifluoromethoxy)phenol are the basic building blocks numerous pharmaceutical drugs.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Quantity | 1g |
Melting Point | 51°C to 55°C |
Formula Weight | 257.01 |
Percent Purity | 98% |
Chemical Name or Material | 4-Bromo-2-(trifluoromethoxy)phenol |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures