Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-2-(trifluoromethyl)anisole, 97%

Click to view available options
Quantity:
2g
10g
1514-11-0
C8H6BrF3O
255.034
MFCD01631579
XTFPWZDSCKENBJ-UHFFFAOYSA-N
4-bromo-2-trifluoromethyl anisole,4-bromo-1-methoxy-2-trifluoromethyl benzene,4-methoxy-3-trifluoromethyl bromobenzene,5-bromo-2-methoxybenzotrifluoride,4-bromo-1-methoxy-2-trifluoromethyl-benzene,2-trifluoromethyl-4-bromoanisole,3-bromo-6-methoxybenzotrifluoride,1-bromo-4-methoxy-3-trifluoromethyl benzene,acmc-20amfg,pubchem4441
2775304
4-bromo-1-methoxy-2-(trifluoromethyl)benzene
COC1=C(C=C(C=C1)Br)C(F)(F)F
1514-11-0
C8H6BrF3O
255.034
MFCD01631579
XTFPWZDSCKENBJ-UHFFFAOYSA-N
4-bromo-2-trifluoromethyl anisole,4-bromo-1-methoxy-2-trifluoromethyl benzene,4-methoxy-3-trifluoromethyl bromobenzene,5-bromo-2-methoxybenzotrifluoride,4-bromo-1-methoxy-2-trifluoromethyl-benzene,2-trifluoromethyl-4-bromoanisole,3-bromo-6-methoxybenzotrifluoride,1-bromo-4-methoxy-3-trifluoromethyl benzene,acmc-20amfg,pubchem4441
2775304
4-bromo-1-methoxy-2-(trifluoromethyl)benzene
COC1=C(C=C(C=C1)Br)C(F)(F)F
Boiling Point 120°C (30mmHg)
Quantity 10g
Beilstein 2525206
Formula Weight 255.04
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-(trifluoromethyl)anisole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning