Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-3,5-dichloropyridine, 95%

Click to view available options
Quantity:
250mg
1g
5g
343781-45-3
C5H2BrCl2N
226.882
MFCD06797978
LOKLQBZBVPYAQO-UHFFFAOYSA-N
3,5-dichloro-4-bromo-pyridine,pubchem21381,4-bromo-3,5-dichloropyridin,3,5-dichloro-4-bromopyridine,pyridine,4-bromo-3,5-dichloro,4-bromanyl-3,5-bis chloranyl pyridine
10680788
4-bromo-3,5-dichloropyridine
C1=C(C(=C(C=N1)Cl)Br)Cl
343781-45-3
C5H2BrCl2N
226.882
MFCD06797978
LOKLQBZBVPYAQO-UHFFFAOYSA-N
3,5-dichloro-4-bromo-pyridine,pubchem21381,4-bromo-3,5-dichloropyridin,3,5-dichloro-4-bromopyridine,pyridine,4-bromo-3,5-dichloro,4-bromanyl-3,5-bis chloranyl pyridine
10680788
4-bromo-3,5-dichloropyridine
C1=C(C(=C(C=N1)Cl)Br)Cl
Quantity 250mg
Color White
Melting Point 75°C to 76°C
UN Number UN2811
Beilstein 8760531
Formula Weight 226.88
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 4-Bromo-3,5-dichloropyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.