Alfa Aesar™ 4-Bromo-3,5-difluorophenol, 99%

Manufacturer: Alfa Aesar™ H2615303

Quantity 1g

 View more versions of this product

Catalog No. AAH2615303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 130191-91-2
    Molecular Formula C6H3BrF2O
    Formula Weight 208.99
    MDL Number MFCD08062385
    Chemical Name or Material 4-Bromo-3,5-difluorophenol

    Specifications

    Formula Weight 208.99
    Molecular Formula C6H3BrF2O
    MDL Number MFCD08062385
    CAS 130191-91-2
    Melting Point 72° to 78°C
    Quantity 1g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 4-Bromo-3,5-difluorophenol
    Assay 99%

    Description

    Applications
    4-Bromo-3,5-difluorophenol is used as intermediates of Liquid Crystals. Reactant in the synthesis of difluoroethyleneoxy group liquid crystal.

    Solubility
    Sparingly Soluble (8.2E-5 g/L) (25°C).

    Notes
    Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

    GHS H Statement
    H301-H311-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit