Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 4-Bromo-3-chlorophenol, 98%
Click to view available options
Quantity:
1g
5g
Description
Applications
4-Bromo-3-chlorophenol is used as a precursor in organic synthesis to prepare 4-(benzyloxy)-l-bromo-2-chlorobenzene by reaction with benzyl bromide.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
4-Bromo-3-chlorophenol is used as a precursor in organic synthesis to prepare 4-(benzyloxy)-l-bromo-2-chlorobenzene by reaction with benzyl bromide.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Odor | Phenol-like |
Melting Point | 64°C to 68°C |
Quantity | 1g |
Beilstein | 3236445 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 207.46 |
Percent Purity | 98% |
Chemical Name or Material | 4-Bromo-3-chlorophenol |
Safety and Handling
GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen