Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 4-Bromo-3-ethoxyaniline hydrochloride, 98%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Quantity | 5g |
Melting Point | 186°C to 189°C |
Formula Weight | 216.08 |
Percent Purity | 98% |
Chemical Name or Material | 4-Bromo-3-ethoxyaniline hydrochloride |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures