Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Alfa Aesar™ 4'-Bromo-3'-fluoroacetanilide, 98+%

Click to view available options
Quantity:
10g
50g
351-30-4
C8H7BrFNO
232.052
MFCD00672937
GAQUWJRFYLETKX-UHFFFAOYSA-N
n-4-bromo-3-fluorophenyl acetamide,4'-bromo-3'-fluoroacetanilide,4-bromo-3-fluoroacetanilide,4-bromo-3'-fluoroacetanilide,acetamide, n-4-bromo-3-fluorophenyl,acmc-1cs5g,acetamide,n-4-bromo-3-fluorophenyl,n-4-bromanyl-3-fluoranyl-phenyl ethanamide
2773316
N-(4-bromo-3-fluorophenyl)acetamide
CC(=O)NC1=CC(=C(C=C1)Br)F
351-30-4
C8H7BrFNO
232.052
MFCD00672937
GAQUWJRFYLETKX-UHFFFAOYSA-N
n-4-bromo-3-fluorophenyl acetamide,4'-bromo-3'-fluoroacetanilide,4-bromo-3-fluoroacetanilide,4-bromo-3'-fluoroacetanilide,acetamide, n-4-bromo-3-fluorophenyl,acmc-1cs5g,acetamide,n-4-bromo-3-fluorophenyl,n-4-bromanyl-3-fluoranyl-phenyl ethanamide
2773316
N-(4-bromo-3-fluorophenyl)acetamide
CC(=O)NC1=CC(=C(C=C1)Br)F
Quantity 50g
Melting Point 149°C to 152°C
Formula Weight 232.05
Percent Purity ≥98%
Chemical Name or Material 4'-Bromo-3'-fluoroacetanilide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning