Alfa Aesar™ 4-Bromo-3-fluoroanisole, 97%

Manufacturer: Alfa Aesar™ H2731903

Quantity 1g

 View more versions of this product

Catalog No. AAH2731903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 458-50-4
    Molecular Formula C7H6BrFO
    Formula Weight 205.03
    MDL Number MFCD01310983
    Synonym 1-Bromo-2-fluoro-4-methoxybenzene
    Chemical Name or Material 4-Bromo-3-fluoroanisole

    Specifications

    Formula Weight 205.03
    Melting Point 217° to 219°C
    Quantity 1g
    Flash Point 85°C (185°F)
    Molecular Formula C7H6BrFO
    MDL Number MFCD01310983
    CAS 458-50-4
    Synonym 1-Bromo-2-fluoro-4-methoxybenzene
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 4-Bromo-3-fluoroanisole
    Assay 97%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit