Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-3-fluorobenzoic acid, 98+%

Click to view available options
Quantity:
1g
5g
153556-42-4
C7H4BrFO2
219.009
MFCD00672932
RMYOGXPGIDWJLU-UHFFFAOYSA-N
3-fluoro-4-bromobenzoic acid,benzoic acid, 4-bromo-3-fluoro,3-fluoro-4-bromo benzoic acid,4-bromo-3-fluoro-benzoic acid,pubchem1316,intermediates-zcf02169,acmc-1c1h6,ksc497g7p,4-bromo-3-fluoro benzoic acid
2773341
4-bromo-3-fluorobenzoic acid
C1=CC(=C(C=C1C(=O)O)F)Br
153556-42-4
C7H4BrFO2
219.009
MFCD00672932
RMYOGXPGIDWJLU-UHFFFAOYSA-N
3-fluoro-4-bromobenzoic acid,benzoic acid, 4-bromo-3-fluoro,3-fluoro-4-bromo benzoic acid,4-bromo-3-fluoro-benzoic acid,pubchem1316,intermediates-zcf02169,acmc-1c1h6,ksc497g7p,4-bromo-3-fluoro benzoic acid
2773341
4-bromo-3-fluorobenzoic acid
C1=CC(=C(C=C1C(=O)O)F)Br
Quantity 5g
Melting Point 210°C to 213°C
Formula Weight 219.01
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-3-fluorobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes