Learn More
eMolecules​ 4-Bromo-3-fluorobenzonitrile | 133059-44-6 | 5G | Purity: 95% DFS Item
Manufacturer:  eMolecules​ 117540647
Combi-Blocks | 4-Bromo-3-fluorobenzonitrile | 5G | 133059-44-6 | MFCD07782069
Catalog No. 50-157-6059
Due to product restrictions, please to purchase or view availability for this product.