Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-3-fluorophenol, 98%

Click to view available options
Quantity:
5g
25g
121219-03-2
C6H4BrFO
190.999
MFCD00051907
MRQYTJXVULSNIS-UHFFFAOYSA-N
3-fluoro-4-bromophenol,4-bromo-3-fluoro-phenol,phenol, 4-bromo-3-fluoro,pubchem1489,4-bromo-3-flourophenol,3-fluoro-4-bromo phenol,3-fluoro-4-bromo-phenol,4-bromo-3-fluoro phenol,4-bromo-5-fluoro-phenol,acmc-209a9m
2779153
4-bromo-3-fluorophenol
C1=CC(=C(C=C1O)F)Br
121219-03-2
C6H4BrFO
190.999
MFCD00051907
MRQYTJXVULSNIS-UHFFFAOYSA-N
3-fluoro-4-bromophenol,4-bromo-3-fluoro-phenol,phenol, 4-bromo-3-fluoro,pubchem1489,4-bromo-3-flourophenol,3-fluoro-4-bromo phenol,3-fluoro-4-bromo-phenol,4-bromo-3-fluoro phenol,4-bromo-5-fluoro-phenol,acmc-209a9m
2779153
4-bromo-3-fluorophenol
C1=CC(=C(C=C1O)F)Br
Melting Point 71°C to 73°C
Quantity 5g
Formula Weight 191
Percent Purity 98%
Chemical Name or Material 4-Bromo-3-fluorophenol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures