Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-3-hydroxybenzyl alcohol, 95%

Click to view available options
Quantity:
250mg
1g
2737-19-1
C7H7BrO2
203.035
MFCD14705103
JYYCVESONPFMEW-UHFFFAOYSA-N
2-bromo-5-hydroxymethyl phenol,4-bromo-3-hydroxybenzyl alcohol,benzenemethanol, 4-bromo-3-hydroxy,4-bromo-3-hydroxybenzyloxamine,2-bromo-5-hydroxymethyl-phenol,4-bromo-3-hydroxy-benzyl alcohol,benzenemethanol,4-bromo-3-hydroxy
13551344
2-bromo-5-(hydroxymethyl)phenol
C1=CC(=C(C=C1CO)O)Br
2737-19-1
C7H7BrO2
203.035
MFCD14705103
JYYCVESONPFMEW-UHFFFAOYSA-N
2-bromo-5-hydroxymethyl phenol,4-bromo-3-hydroxybenzyl alcohol,benzenemethanol, 4-bromo-3-hydroxy,4-bromo-3-hydroxybenzyloxamine,2-bromo-5-hydroxymethyl-phenol,4-bromo-3-hydroxy-benzyl alcohol,benzenemethanol,4-bromo-3-hydroxy
13551344
2-bromo-5-(hydroxymethyl)phenol
C1=CC(=C(C=C1CO)O)Br
Quantity 1g
Beilstein 1936254
Formula Weight 203.04
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 4-Bromo-3-hydroxybenzyl alcohol