Alfa Aesar™ 4'-Bromo-3'-methylacetanilide, 98%

Manufacturer: Alfa Aesar™ B2198014

Quantity 25g

 View more versions of this product

Catalog No. AAB2198014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 90914-81-1
    Molecular Formula C9H10BrNO
    Formula Weight 228.09
    MDL Number MFCD00040822
    Chemical Name or Material 4'-Bromo-3'-methylacetanilide

    Specifications

    Formula Weight 228.09
    Molecular Formula C9H10BrNO
    MDL Number MFCD00040822
    CAS 90914-81-1
    Melting Point 102° to 104°C
    Quantity 25g
    Chemical Name or Material 4'-Bromo-3'-methylacetanilide
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit