Alfa Aesar™ 4-Bromo-3-methylanisole, 97%

Manufacturer: Alfa Aesar™ A1231818

Quantity 50g

 View more versions of this product

Catalog No. AAA1231818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 27060-75-9
    Molecular Formula C8H9BrO
    Formula Weight 201.06
    MDL Number MFCD00060687
    Beilstein 1933865
    Synonym 2-Bromo-5-methoxytoluene
    Chemical Name or Material 4-Bromo-3-methylanisole

    Specifications

    Formula Weight 201.06
    TSCA TSCA
    CAS 27060-75-9
    Synonym 2-Bromo-5-methoxytoluene
    MDL Number MFCD00060687
    Quantity 50g
    Molecular Formula C8H9BrO
    Beilstein 1933865
    Boiling Point 111° to 112°C (15mmHg)
    Flash Point >110°C (230°F)
    Chemical Name or Material 4-Bromo-3-methylanisole
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit