Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-3-methylbenzenesulfonyl chloride, 97%

Click to view available options
Quantity:
1g
5g
72256-93-0
C7H6BrClO2S
269.537
MFCD03094696
JRNDXUKMWFVEIC-UHFFFAOYSA-N
4-bromo-3-methylbenzene-1-sulfonyl chloride,4-bromo-3-methylbenzenesulphonyl chloride,4-bromo-3-methylbenzenesulphonylchloride,4-bromo-3-methyl-benzenesulfonyl chloride,6-bromo-m-toluene-3-sulfonyl chloride,pubchem15805,acmc-1bho4,jrndxukmwfveic-uhfffaoysa,4-bromo-3-methylphenyl chlorosulfone,4-bromo-3-methylbenzene sulfonyl chloride
2737503
4-bromo-3-methylbenzenesulfonyl chloride
CC1=C(C=CC(=C1)S(=O)(=O)Cl)Br
72256-93-0
C7H6BrClO2S
269.537
MFCD03094696
JRNDXUKMWFVEIC-UHFFFAOYSA-N
4-bromo-3-methylbenzene-1-sulfonyl chloride,4-bromo-3-methylbenzenesulphonyl chloride,4-bromo-3-methylbenzenesulphonylchloride,4-bromo-3-methyl-benzenesulfonyl chloride,6-bromo-m-toluene-3-sulfonyl chloride,pubchem15805,acmc-1bho4,jrndxukmwfveic-uhfffaoysa,4-bromo-3-methylphenyl chlorosulfone,4-bromo-3-methylbenzene sulfonyl chloride
2737503
4-bromo-3-methylbenzenesulfonyl chloride
CC1=C(C=CC(=C1)S(=O)(=O)Cl)Br
Melting Point 57°C to 61°C
Quantity 1g
UN Number UN3261
Beilstein 3254604
Formula Weight 269.55
Percent Purity 97%
Chemical Name or Material 4-Bromo-3-methylbenzenesulfonyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.