Alfa Aesar™ 4-Bromo-3-methylbenzonitrile, 98%

Manufacturer: Alfa Aesar™ B2537703

Quantity 1g

 View more versions of this product

Catalog No. AAB2537703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 41963-20-6
    Molecular Formula C8H6BrN
    Formula Weight 196.05
    MDL Number MFCD00031538
    UN Number UN3439
    Beilstein 2354972
    Chemical Name or Material 4-Bromo-3-methylbenzonitrile

    Specifications

    Formula Weight 196.05
    Molecular Formula C8H6BrN
    MDL Number MFCD00031538
    Melting Point 52° to 54°C
    Quantity 1g
    CAS 41963-20-6
    UN Number UN3439
    Beilstein 2354972
    Boiling Point 129° to 130°C (5mmHg)
    Flash Point 96°C (205°F)
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Health Hazard 1 Danger
    Chemical Name or Material 4-Bromo-3-methylbenzonitrile
    Assay 98%

    Description

    Applications
    4-bromo-3-methylbenzenecarbonitrile is a nitrile compound for proteomics research & a pharmaceutical intermediate.

    Solubility
    Insoluble in water.

    Notes
    Store in refrigerator at (2-4°C). Incompatible material is strong oxidizing agents.

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit