Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-3-methylphenol, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
14472-14-1
C7H7BrO
187.04
MFCD00079723
GPOQODYGMUTOQL-UHFFFAOYSA-N
4-bromo-3-methyl-phenol,4-bromo-m-cresol,phenol, 4-bromo-3-methyl,4-bromo-3-methyl phenol,4-bromo-5-methyl-phenol,4-brom-3-methylphenol,zlchem 434,pubchem2201,3-methyl-4-bromophenol,2-bromo-5-hydroxytoluene
72857
4-bromo-3-methylphenol
CC1=C(C=CC(=C1)O)Br
14472-14-1
C7H7BrO
187.04
MFCD00079723
GPOQODYGMUTOQL-UHFFFAOYSA-N
4-bromo-3-methyl-phenol,4-bromo-m-cresol,phenol, 4-bromo-3-methyl,4-bromo-3-methyl phenol,4-bromo-5-methyl-phenol,4-brom-3-methylphenol,zlchem 434,pubchem2201,3-methyl-4-bromophenol,2-bromo-5-hydroxytoluene
72857
4-bromo-3-methylphenol
CC1=C(C=CC(=C1)O)Br
Color White
Boiling Point 142°C to 145°C (23.0mmHg)
Melting Point 57°C to 61°C
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula BrC6H3(CH3)OH
Beilstein 06, IV, 1321
Formula Weight 187.04
Physical Form Crystalline Powder or Chunks
Percent Purity 98%
Chemical Name or Material 4-Bromo-3-methylphenol, 98%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 238-464-3