Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Bromo-3'-nitroacetophenone 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
18640-58-9
C8H6BrNO3
244.044
MFCD00016985
YFVOFFKNHQTQQE-UHFFFAOYSA-N
1-4-bromo-3-nitrophenyl ethanone,4'-bromo-3'-nitroacetophenone,1-4-bromo-3-nitrophenyl ethan-1-one,4-bromo-3-nitroacetophenone,4-bromo-3'-nitroacetophenone,ethanone, 1-4-bromo-3-nitrophenyl,acmc-209eny,3-nitro-4-bromoacetophenone,4-bromo-3-nitro acetophenone,3'-nitro-4'-bromoacetophenone
87737
1-(4-bromo-3-nitrophenyl)ethanone
CC(=O)C1=CC(=C(C=C1)Br)[N+](=O)[O-]
18640-58-9
C8H6BrNO3
244.044
MFCD00016985
YFVOFFKNHQTQQE-UHFFFAOYSA-N
1-4-bromo-3-nitrophenyl ethanone,4'-bromo-3'-nitroacetophenone,1-4-bromo-3-nitrophenyl ethan-1-one,4-bromo-3-nitroacetophenone,4-bromo-3'-nitroacetophenone,ethanone, 1-4-bromo-3-nitrophenyl,acmc-209eny,3-nitro-4-bromoacetophenone,4-bromo-3-nitro acetophenone,3'-nitro-4'-bromoacetophenone
87737
1-(4-bromo-3-nitrophenyl)ethanone
CC(=O)C1=CC(=C(C=C1)Br)[N+](=O)[O-]
Quantity 5g
Color White to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 118.0°C to 120.0°C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula Br(NO2)C6H3COCH3
Formula Weight 244.04
Physical Form Amorphous Powder
Percent Purity ≥99%
Chemical Name or Material 4'-Bromo-3'-nitroacetophenone, 99+%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LACHRYMATOR