Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-3-nitrotoluene, 98+%

Click to view available options
Quantity:
25g
100g
5326-34-1
C7H6BrNO2
216.03
MFCD00024180
UPBUTKQMDPHQAQ-UHFFFAOYSA-N
4-bromo-3-nitrotoluene, 2-bromo-5-methylnitrobenzene, benzene, 1-bromo-4-methyl-2-nitro, bromonitrotoluene-4, 1-bromo-4-methyl-2-nitro-benzene, 3-nitro-4-bromotoluene, pubchem9585, acmc-209l5d, ksc490m7t, tpc-i121
79224
1-bromo-4-methyl-2-nitrobenzene
CC1=CC=C(Br)C(=C1)[N+]([O-])=O
5326-34-1
C7H6BrNO2
216.03
MFCD00024180
UPBUTKQMDPHQAQ-UHFFFAOYSA-N
4-bromo-3-nitrotoluene, 2-bromo-5-methylnitrobenzene, benzene, 1-bromo-4-methyl-2-nitro, bromonitrotoluene-4, 1-bromo-4-methyl-2-nitro-benzene, 3-nitro-4-bromotoluene, pubchem9585, acmc-209l5d, ksc490m7t, tpc-i121
79224
1-bromo-4-methyl-2-nitrobenzene
CC1=CC=C(Br)C(=C1)[N+]([O-])=O
Sensitivity Light sensitive
Density 1.578
Quantity 25g
Melting Point 29°C to 32°C
Boiling Point 96°C (1mmHg)
Flash Point >110°C (230°F)
Beilstein 1945925
Formula Weight 216.04
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-3-nitrotoluene

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 226-203-6

TSCA : No

Recommended Storage : Ambient temperatures