Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-3-(trifluoromethyl)-1H-pyrazole, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
19968-17-3
C4H2BrF3N2
214.97
JTHNMRUVJDWVMJ-UHFFFAOYSA-N
4-bromo-3-trifluoromethyl-1h-pyrazole,4-bromo-5-trifluoromethyl-1h-pyrazole,1h-pyrazole, 4-bromo-3-trifluoromethyl,1h-pyrazole, 4-bromo-5-trifluoromethyl,1h-pyrazole,4-bromo-3-trifluoromethyl,4-bromo-3-trifluoromethyl pyrazole,4-bromo-3-trifluoromethyl-2h-pyrazole,acmc-209f4m,4-bromo-3 trifluoromethyl-1h-pyrazole
7147398
4-bromo-5-(trifluoromethyl)-1H-pyrazole
C1=NNC(=C1Br)C(F)(F)F
19968-17-3
C4H2BrF3N2
214.97
JTHNMRUVJDWVMJ-UHFFFAOYSA-N
4-bromo-3-trifluoromethyl-1h-pyrazole,4-bromo-5-trifluoromethyl-1h-pyrazole,1h-pyrazole, 4-bromo-3-trifluoromethyl,1h-pyrazole, 4-bromo-5-trifluoromethyl,1h-pyrazole,4-bromo-3-trifluoromethyl,4-bromo-3-trifluoromethyl pyrazole,4-bromo-3-trifluoromethyl-2h-pyrazole,acmc-209f4m,4-bromo-3 trifluoromethyl-1h-pyrazole
7147398
4-bromo-5-(trifluoromethyl)-1H-pyrazole
C1=NNC(=C1Br)C(F)(F)F
Packaging Glass Bottle
Melting Point 112°C to 114°C
Quantity 1g
Color White
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 214.97
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Bromo-3-(trifluoromethyl)-1H-pyrazole
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning