Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4'-Bromo-3'-(trifluoromethyl)acetanilide, 97%

Click to view available options
Quantity:
2.5g
10g
41513-05-7
C9H7BrF3NO
282.06
MFCD00052361
AOHSBSXIGFIMNC-UHFFFAOYSA-N
n-4-bromo-3-trifluoromethyl phenyl acetamide,5-acetamido-2-bromobenzotrifluoride,4-bromo-3-trifluoromethyl acetanilide,4'-bromo-3'-trifluoromethyl acetanilide,n-4-bromo-3-trifluoromethyl phenyl acet amide,n1-4-bromo-3-trifluoromethyl phenyl acetamide,pubchem2757,maybridge4_003018,buttpark 37\11-74
697838
N-[4-bromo-3-(trifluoromethyl)phenyl]acetamide
CC(=O)NC1=CC(=C(C=C1)Br)C(F)(F)F
41513-05-7
C9H7BrF3NO
282.06
MFCD00052361
AOHSBSXIGFIMNC-UHFFFAOYSA-N
n-4-bromo-3-trifluoromethyl phenyl acetamide,5-acetamido-2-bromobenzotrifluoride,4-bromo-3-trifluoromethyl acetanilide,4'-bromo-3'-trifluoromethyl acetanilide,n-4-bromo-3-trifluoromethyl phenyl acet amide,n1-4-bromo-3-trifluoromethyl phenyl acetamide,pubchem2757,maybridge4_003018,buttpark 37\11-74
697838
N-[4-bromo-3-(trifluoromethyl)phenyl]acetamide
CC(=O)NC1=CC(=C(C=C1)Br)C(F)(F)F
Quantity 2.5g
Melting Point 116°C to 119°C
Formula Weight 282.07
Percent Purity 97%
Chemical Name or Material 4'-Bromo-3'-(trifluoromethyl)acetanilide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures