Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-3-(trifluoromethyl)phenol, 98%

Click to view available options
Quantity:
5g
25g
320-49-0
C7H4BrF3O
241.007
MFCD03095342
VOWPIDJSINRFPZ-UHFFFAOYSA-N
4-bromo-3-trifluoromethyl phenol,2-bromo-5-hydroxybenzotrifluoride,3-trifluoromethyl-4-bromophenol,phenol, 4-bromo-3-trifluoromethyl,4-bromo-3-trifluromethylphenol,4-bromo-3-trifluoromethyl-phenol,btfp,pubchem2111,acmc-1ahmz,3-trifluoromethyl-4-bromo phenol
22277307
4-bromo-3-(trifluoromethyl)phenol
C1=CC(=C(C=C1O)C(F)(F)F)Br
320-49-0
C7H4BrF3O
241.007
MFCD03095342
VOWPIDJSINRFPZ-UHFFFAOYSA-N
4-bromo-3-trifluoromethyl phenol,2-bromo-5-hydroxybenzotrifluoride,3-trifluoromethyl-4-bromophenol,phenol, 4-bromo-3-trifluoromethyl,4-bromo-3-trifluromethylphenol,4-bromo-3-trifluoromethyl-phenol,btfp,pubchem2111,acmc-1ahmz,3-trifluoromethyl-4-bromo phenol
22277307
4-bromo-3-(trifluoromethyl)phenol
C1=CC(=C(C=C1O)C(F)(F)F)Br
Color Yellow
Quantity 25g
Melting Point 44°C to 46°C
UN Number UN3261
Formula Weight 241.01
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-Bromo-3-(trifluoromethyl)phenol

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.