Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-4'-iodobiphenyl, 98%

Click to view available options
Quantity:
25g
100g
105946-82-5
C12H8BrI
359.004
MFCD17676204
GWOAJJWBCSUGHH-UHFFFAOYSA-N
4-bromo-4'-iodobiphenyl,4-bromo-4'-iodo-1,1'-biphenyl,1,1'-biphenyl, 4-bromo-4'-iodo,1-bromo-4-4-iodophenyl benzene,4'-bromo-4-iodobiphenyl,4-iodo-4'-bromobiphenyl,4-bromo-4'-iodo-biphenyl,4-bromo-4/'-iodobiphenyl,ksc164g2f,4-bromo-4'-iodobiphenyl
10904452
1-bromo-4-(4-iodophenyl)benzene
C1=CC(=CC=C1C2=CC=C(C=C2)I)Br
105946-82-5
C12H8BrI
359.004
MFCD17676204
GWOAJJWBCSUGHH-UHFFFAOYSA-N
4-bromo-4'-iodobiphenyl,4-bromo-4'-iodo-1,1'-biphenyl,1,1'-biphenyl, 4-bromo-4'-iodo,1-bromo-4-4-iodophenyl benzene,4'-bromo-4-iodobiphenyl,4-iodo-4'-bromobiphenyl,4-bromo-4'-iodo-biphenyl,4-bromo-4/'-iodobiphenyl,ksc164g2f,4-bromo-4'-iodobiphenyl
10904452
1-bromo-4-(4-iodophenyl)benzene
C1=CC(=CC=C1C2=CC=C(C=C2)I)Br
Melting Point 175°C to 179°C (literature)
Sensitivity Light Sensitive
Quantity 25g
UN Number UN3152
Beilstein 2518914
Formula Weight 359
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-Bromo-4'-iodobiphenyl

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packaging Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, SOLID