Alfa Aesar™ 4-Bromo-4'-methylbiphenyl, 97%

Manufacturer: Alfa Aesar™ H50596MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50596MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 50670-49-0
    Molecular Formula C13H11Br
    Formula Weight 247.14
    MDL Number MFCD00466236
    Chemical Name or Material 4-Bromo-4'-methylbiphenyl

    Specifications

    Formula Weight 247.14
    Molecular Formula C13H11Br
    MDL Number MFCD00466236
    CAS 50670-49-0
    Melting Point 136°C
    Quantity 250mg
    Chemical Name or Material 4-Bromo-4'-methylbiphenyl
    Assay 97%

    Description

    Applications
    4-Bromo-4 ft.-methylbiphenyl is used as pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit