Alfa Aesar™ 4-Bromo-5-nonanone, 98%

Manufacturer: Alfa Aesar™ H3213603

Quantity 1g

 View more versions of this product

Catalog No. AAH3213603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 42330-11-0
    Molecular Formula C9H17BrO
    Formula Weight 221.14
    MDL Number MFCD09757527
    UN Number UN3265
    Chemical Name or Material 4-Bromo-5-nonanone

    Specifications

    Formula Weight 221.14
    Boiling Point 62°C (1mmHg)
    Quantity 1g
    Flash Point 92°C (198°F)
    Molecular Formula C9H17BrO
    MDL Number MFCD09757527
    CAS 42330-11-0
    UN Number UN3265
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Health Hazard 1 Danger
    Chemical Name or Material 4-Bromo-5-nonanone
    Assay 98%

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit