Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-7-azaindole, 95%

Click to view available options
Quantity:
1g
5g
348640-06-2
C7H5BrN2
197.035
MFCD08272233
LEZHTYOQWQEBLH-UHFFFAOYSA-N
4-bromo-7-azaindole,4-bromo-1h-pyrrolo 2,3-b pyridine,4-bromo-1h-pyrrolo 2,3-b pyridine,1h-pyrrolo 2,3-b pyridine, 4-bromo,4-bromo-7-azaindol,4-bromo-1h-pyrrolo-2,3 pyridine,4-bromo-1h-pyrrolo 2,3-b pyridine,pubchem14706,4-bromo-7-aza-1h-indole,ksc497k7l
22273643
4-bromo-1H-pyrrolo[2,3-b]pyridine
C1=CNC2=NC=CC(=C21)Br
348640-06-2
C7H5BrN2
197.035
MFCD08272233
LEZHTYOQWQEBLH-UHFFFAOYSA-N
4-bromo-7-azaindole,4-bromo-1h-pyrrolo 2,3-b pyridine,4-bromo-1h-pyrrolo 2,3-b pyridine,1h-pyrrolo 2,3-b pyridine, 4-bromo,4-bromo-7-azaindol,4-bromo-1h-pyrrolo-2,3 pyridine,4-bromo-1h-pyrrolo 2,3-b pyridine,pubchem14706,4-bromo-7-aza-1h-indole,ksc497k7l
22273643
4-bromo-1H-pyrrolo[2,3-b]pyridine
C1=CNC2=NC=CC(=C21)Br
Quantity 5g
Color Yellow to Orange
Melting Point 178°C to 183°C
UN Number UN2811
Beilstein 9695211
Formula Weight 197.04
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 4-Bromo-7-azaindole

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.