Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-9,9-dimethylfluorene, 98%

Click to view available options
Quantity:
250mg
1g
5g
942615-32-9
C15H13Br
273.173
MFCD23099415
SXOUNESKHJIXNK-UHFFFAOYSA-N
4-bromo-9,9-dimethyl-9h-fluorene,4-bromo-9,9-dimethyl fluorene,4-bromo-9,9-dimethyl-fluorene
59182836
4-bromo-9,9-dimethylfluorene
CC1(C2=C(C3=CC=CC=C31)C(=CC=C2)Br)C
942615-32-9
C15H13Br
273.173
MFCD23099415
SXOUNESKHJIXNK-UHFFFAOYSA-N
4-bromo-9,9-dimethyl-9h-fluorene,4-bromo-9,9-dimethyl fluorene,4-bromo-9,9-dimethyl-fluorene
59182836
4-bromo-9,9-dimethylfluorene
CC1(C2=C(C3=CC=CC=C31)C(=CC=C2)Br)C
Quantity 250mg
Beilstein 22540802
Formula Weight 273.17
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-Bromo-9,9-dimethylfluorene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning