Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-D-phenylalanine, 95%

Click to view available options
Quantity:
250mg
1g
5g
62561-74-4
C9H10BrNO2
244.088
MFCD01075126
PEMUHKUIQHFMTH-MRVPVSSYSA-N
4-bromo-d-phenylalanine,h-d-phe 4-br-oh,d-4-bromophenylalanine,r-2-amino-3-4-bromophenyl propanoic acid,2r-2-amino-3-4-bromophenyl propanoic acid,d-4-br-phe-oh,4-bromo-d-phe-oh,d-phenylalanine, 4-bromo,h-p-bromo-d-phe-oh,d-phe 4-br-oh
671215
(2R)-2-amino-3-(4-bromophenyl)propanoic acid
C1=CC(=CC=C1CC(C(=O)O)N)Br
62561-74-4
C9H10BrNO2
244.088
MFCD01075126
PEMUHKUIQHFMTH-MRVPVSSYSA-N
4-bromo-d-phenylalanine,h-d-phe 4-br-oh,d-4-bromophenylalanine,r-2-amino-3-4-bromophenyl propanoic acid,2r-2-amino-3-4-bromophenyl propanoic acid,d-4-br-phe-oh,4-bromo-d-phe-oh,d-phenylalanine, 4-bromo,h-p-bromo-d-phe-oh,d-phe 4-br-oh
671215
(2R)-2-amino-3-(4-bromophenyl)propanoic acid
C1=CC(=CC=C1CC(C(=O)O)N)Br
Formula Weight 244.09
Percent Purity 95%
Quantity 1g
Chemical Name or Material 4-Bromo-D-phenylalanine

TSCA : No

Recommended Storage : Ambient temperatures