Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-N-Fmoc-D-phenylalanine, 95%

Click to view available options
Quantity:
1g
5g
198545-76-5
C24H20BrNO4
466.331
MFCD00273459
TVBAVBWXRDHONF-JOCHJYFZSA-N
fmoc-4-bromo-d-phenylalanine,r-n-fmoc-4-bromophenylalanine,fmoc-d-4-bromophenylalanine,fmoc-d-phe 4-br-oh,fmoc-d-4-br-phe-oh,fmoc-4'-bromo-d-phe,fmoc-d-4-bromophe,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-bromophenyl propanoic acid,n-9-fluorenylmethoxycarbonyl-4-bromophenyl-d-alanine,n-alpha-9-fluorenylmethyloxycarbonyl-4-bromo-d-phenylalanine
7020431
(2R)-3-(4-bromophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Br)C(=O)O
198545-76-5
C24H20BrNO4
466.331
MFCD00273459
TVBAVBWXRDHONF-JOCHJYFZSA-N
fmoc-4-bromo-d-phenylalanine,r-n-fmoc-4-bromophenylalanine,fmoc-d-4-bromophenylalanine,fmoc-d-phe 4-br-oh,fmoc-d-4-br-phe-oh,fmoc-4'-bromo-d-phe,fmoc-d-4-bromophe,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-bromophenyl propanoic acid,n-9-fluorenylmethoxycarbonyl-4-bromophenyl-d-alanine,n-alpha-9-fluorenylmethyloxycarbonyl-4-bromo-d-phenylalanine
7020431
(2R)-3-(4-bromophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Br)C(=O)O
Formula Weight 426.47
Physical Form Solid
Percent Purity 95%
Quantity 5g
Chemical Name or Material 4-Bromo-N-Fmoc-D-phenylalanine

storageNote1 : -30° to -10°C