Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-N-methylaniline, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
6911-87-1
C7H8BrN
186.05
MFCD03030391
AYVPVDWQZAAZCM-UHFFFAOYSA-N
4-bromo-n-methylbenzenamine,benzenamine, 4-bromo-n-methyl,n-methyl-4-bromoaniline,benzenamine,4-bromo-n-methyl,4bromo-n-methylaniline,4-bromo-n-methyl-aniline,n-methyl-4-bromo-aniline,acmc-1cfc7,4-bromanyl-n-methyl-aniline,4-bromo-phenyl-methyl-amine
2757052
4-bromo-N-methylaniline
CNC1=CC=C(C=C1)Br
6911-87-1
C7H8BrN
186.05
MFCD03030391
AYVPVDWQZAAZCM-UHFFFAOYSA-N
4-bromo-n-methylbenzenamine,benzenamine, 4-bromo-n-methyl,n-methyl-4-bromoaniline,benzenamine,4-bromo-n-methyl,4bromo-n-methylaniline,4-bromo-n-methyl-aniline,n-methyl-4-bromo-aniline,acmc-1cfc7,4-bromanyl-n-methyl-aniline,4-bromo-phenyl-methyl-amine
2757052
4-bromo-N-methylaniline
CNC1=CC=C(C=C1)Br
Boiling Point 123°C to 125°C (7.0mmHg)
Refractive Index 1.6120 to 1.614
Quantity 1g
Infrared Spectrum Authentic
Color Brown-Purple to Colorless
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula BrC6H4NHCH3
Formula Weight 186.05
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 4-Bromo-N-methylaniline
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning