Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromo-N,N-dimethylaniline, 98+%

Manufacturer:  Alfa Aesar™ A1085422

Quantity 100g

 View more versions of this product

Catalog No. AAA1085422


  Check Availability
Add to cart

Specifications

Description

Applications
4-Bromo-N,N-dimethylaniline is used as an internal standard in the determination of iodine present as iodide (as in pharmaceuticals), iodate (as in iodized table salt) and covalently bound to organic compounds (as in milk and vegetables).

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, acids and metals. Store at 4°C.
Description & Specifications

Chemical Identifiers

586-77-6
200.08
2206155
C8H10BrN
MFCD00000093
4-Bromo-N,N-dimethylaniline

Specifications

586-77-6
200.08
52° to 56°C
263° to 265°C
>110°C (230°F)
≥98%
C8H10BrN
MFCD00000093
2206155
100g
4-Bromo-N,N-dimethylaniline

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : BW9300000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit