Learn More
4-Bromo-N,N-dimethylaniline, 98+%, Thermo Scientific Chemicals
Description
4-Bromo-N,N-dimethylaniline is used as an internal standard in the determination of iodine present as iodide (as in pharmaceuticals), iodate (as in iodized table salt) and covalently bound to organic compounds (as in milk and vegetables).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Bromo-N,N-dimethylaniline is used as an internal standard in the determination of iodine present as iodide (as in pharmaceuticals), iodate (as in iodized table salt) and covalently bound to organic compounds (as in milk and vegetables).
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, acids and metals. Store at 4°C.
Specifications
Melting Point | 52°C to 56°C |
Boiling Point | 263°C to 265°C |
Flash Point | >110°C (230°F) |
Quantity | 25 g |
Beilstein | 2206155 |
Solubility Information | Soluble in methanol. Insoluble in water. |
Formula Weight | 200.08 |
Percent Purity | ≥98% |
Chemical Name or Material | 4-Bromo-N,N-dimethylaniline |
Safety and Handling
Hazard Category | H302+H312+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H301-H311-H332-H315-H319 Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Hazard Class: 6.1; Packaging Group: III |
EINECSNumber | 209-582-2 |
RTECSNumber | BW9300000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.