Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Bromoacetanilide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
103-88-8
C8H8BrNO
214.06
MFCD00000092
MSLICLMCQYQNPK-UHFFFAOYSA-N
4'-bromoacetanilide,n-4-bromophenyl acetamide,p-bromoacetanilide,4-bromoacetanilide,bromoanilide,acetamide, n-4-bromophenyl,antisepsin,bromoantifebrin,asepsin,p-bromo-n-acetanilide
7683
N-(4-bromophenyl)acetamide
CC(=O)NC1=CC=C(C=C1)Br
103-88-8
C8H8BrNO
214.06
MFCD00000092
MSLICLMCQYQNPK-UHFFFAOYSA-N
4'-bromoacetanilide,n-4-bromophenyl acetamide,p-bromoacetanilide,4-bromoacetanilide,bromoanilide,acetamide, n-4-bromophenyl,antisepsin,bromoantifebrin,asepsin,p-bromo-n-acetanilide
7683
N-(4-bromophenyl)acetamide
CC(=O)NC1=CC=C(C=C1)Br
Quantity 100g
Melting Point 166.0°C to 170.0°C
Color Beige to White or Gray
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3CONHC6H4Br
Beilstein 12,642
Merck Index 15,1406
Solubility Information Solubility in water: practically insoluble. Other solubilities: sparingly soluble in hot water,soluble in benzene,chloroform and ethyl acetate,moderately soluble in alcohol
Formula Weight 214.06
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Chemical Name or Material 4′-Bromoacetanilide
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-154-9