Alfa Aesar™ 4-(Bromoacetyl)benzonitrile, 97%

Manufacturer: Alfa Aesar™ B2538214

Quantity 25g

 View more versions of this product

Catalog No. AAB2538214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20099-89-2
    Molecular Formula C9H6BrNO
    Formula Weight 224.06
    MDL Number MFCD00052931
    UN Number UN2923
    Beilstein 2087648
    Synonym 2-Bromo-4'-cyanoacetophenone; 4-Cyanophenacyl bromide
    Chemical Name or Material 4-(Bromoacetyl)benzonitrile

    Specifications

    Formula Weight 224.06
    Molecular Formula C9H6BrNO
    MDL Number MFCD00052931
    Synonym 2-Bromo-4'-cyanoacetophenone; 4-Cyanophenacyl bromide
    Quantity 25g
    Odor Irritating
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H331-H314-H318
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    CAS 20099-89-2
    UN Number UN2923
    Melting Point 91° to 96°C
    Beilstein 2087648
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 4-(Bromoacetyl)benzonitrile
    Assay 97%

    GHS H Statement
    H301-H311-H331-H314-H318
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit