4-Bromoaniline, 98+%

Manufacturer: Alfa Aesar A1512618

Quantity 50g

 View more versions of this product

Catalog No. AAA1512618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 106-40-1
    Molecular Formula C6H6BrN
    Formula Weight 172.03
    MDL Number MFCD00007822
    UN Number UN2811
    Beilstein 742031
    Merck Index 14,1404
    Chemical Name or Material 4-Bromoaniline

    Specifications

    Formula Weight 172.03
    Quantity 50g
    TSCA TSCA
    MDL Number MFCD00007822
    UN Number UN2811
    Beilstein 742031
    Merck Index 14,1404
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H373-H302-H332-H315-H319
    Toxic in contact with skin.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes seriou
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310-P302+P352
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number BW9280000
    Assay Percent Range ≥98%
    Molecular Formula C6H6BrN
    CAS 106-40-1
    Chemical Name or Material 4-Bromoaniline
    Assay ≥98%

    Description

    Applications
    4-Bromoaniline is used as a precursor in the production of azo dyes and dihydroquinazolines. It is also used as carbon and nitrogen supplement in the culture medium of Moraxella sp. strain G. Further, it is employed in the preparation of para-bromobiphenyl through Gomberg-Bachmann reaction. In addition to this, it is used as a building block in the preparation or pharmaceutical and organic compounds.

    Solubility
    Soluble in alcohol and ether. Insoluble in cold water.

    Notes
    Store in a cool place. Light sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.

    GHS H Statement
    H311-H373-H302-H332-H315-H319
    Toxic in contact with skin.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes skin irritation.
    Causes seriou

    GHS P Statement
    P280h-P305+P351+P338-P309-P310-P302+P352
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit