Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromoaniline, 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
106-40-1
C6H6BrN
172.02
WDFQBORIUYODSI-UHFFFAOYSA-N
p-bromoaniline,4-bromobenzenamine,benzenamine, 4-bromo,p-bromophenylamine,aniline, p-bromo,4-bromo-aniline,4-bromobenzeneamine,p-bromo aniline,4-bromanilinu,4-bromo-phenylamine
7807
4-bromoaniline
C1=CC(=CC=C1N)Br
106-40-1
C6H6BrN
172.02
WDFQBORIUYODSI-UHFFFAOYSA-N
p-bromoaniline,4-bromobenzenamine,benzenamine, 4-bromo,p-bromophenylamine,aniline, p-bromo,4-bromo-aniline,4-bromobenzeneamine,p-bromo aniline,4-bromanilinu,4-bromo-phenylamine
7807
4-bromoaniline
C1=CC(=CC=C1N)Br
Infrared Spectrum Authentic
Packaging Glass bottle
Density 1.4970g/mL
Boiling Point 230.0°C to 250.0°C
Quantity 500g
Flash Point >110°C
Color Brown-Gray to White
Melting Point 60.0°C to 64.0°C
Specific Gravity 1.497
Assay Percent Range 99% min. (GC)
Linear Formula BrC6H4NH2
Beilstein 12,636
Merck Index 15,1411
Solubility Information Solubility in water: < 1 g / l. Other solubilities: soluble in most organic solvents except cold,petroleum,very soluble in alcohol and ether
Formula Weight 172.02
Physical Form Crystalline Powder
Percent Purity 99+%
Chemical Name or Material 4-Bromoaniline
Show More Show Less

GHS H Statement
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 203-393-9