Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromoanisole, 99%, Thermo Scientific Chemicals

Catalog Number AAA1182422
Click to view available options
Quantity:
100 g
500 g
2500 g
104-92-7
C7H7BrO
187.036
MFCD00000097
QJPJQTDYNZXKQF-UHFFFAOYSA-N
4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide
7730
CHEBI:47257
1-bromo-4-methoxybenzene
COC1=CC=C(C=C1)Br
104-92-7
C7H7BrO
187.036
MFCD00000097
QJPJQTDYNZXKQF-UHFFFAOYSA-N
4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide
7730
CHEBI:47257
1-bromo-4-methoxybenzene
COC1=CC=C(C=C1)Br

4-Bromoanisole finds application as an intermediate in synthetic chemistry. It is used in the preparation of aryl 1,3-diketones and ethyl 4-methoxycinnamate. It is a used as a brominating reagent. Further, it is used in Suzuki coupling reaction with phenylboronic acid as well as in Heck reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoanisole finds application as an intermediate in synthetic chemistry. It is used in the preparation of aryl 1,3-diketones and ethyl 4-methoxycinnamate. It is a used as a brominating reagent. Further, it is used in Suzuki coupling reaction with phenylboronic acid as well as in Heck reaction.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY
Melting Point 10°C to 13°C
Density 1.494
Boiling Point 98°C to 99°C (10 mm)
Flash Point 94°C (201°F)
Assay Percent Range 99%
Refractive Index 1.564
Quantity 100 g
Beilstein 1237590
Merck Index 14,1427
Solubility Information Immiscible with water.
Formula Weight 187.04
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Bromoanisole
Show More Show Less
EINECSNumber 203-252-1
RTECSNumber BZ8501000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit