Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromobenzaldehyde diethyl acetal, 98%

Manufacturer:  Alfa Aesar™ B2516322

Quantity 100g

 View more versions of this product

Catalog No. AAB2516322


  Check Availability
Add to cart

Specifications

Description

Applications
4-Bromobenzaldehyde diethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Description & Specifications

Chemical Identifiers

34421-94-8
259.14
2448775
4-Bromobenzaldehyde diethyl acetal
C11H15BrO2
MFCD01863514
4-Bromo-alpha,alpha-diethoxytoluene

Specifications

34421-94-8
259.14
2448775
99° to 101°C (1mmHg)
104°C (219°F)
98%
C11H15BrO2
MFCD01863514
4-Bromo-alpha,alpha-diethoxytoluene
100g
4-Bromobenzaldehyde diethyl acetal
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit