Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromobenzaldehyde diethyl acetal, 99%, Acros Organics

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
34421-94-8
C11H15BrO2
259.14
MFCD01863514
BFSNEBVTOODGHZ-UHFFFAOYSA-N
4-bromobenzaldehyde diethyl acetal,1-bromo-4-diethoxymethyl benzene,4-bromobenzaldehydediethylacetal,benzene, 1-bromo-4-diethoxymethyl,4-bromophenyl diethoxymethane,4-bromobenzaldehyde diethylacetal,4-bromo-alpha,alpha-diethoxytoluene,pubchem5372,acmc-1aigt,4-bromobenzaldehydediethyl acetal
688340
1-bromo-4-(diethoxymethyl)benzene
CCOC(C1=CC=C(C=C1)Br)OCC
34421-94-8
C11H15BrO2
259.14
MFCD01863514
BFSNEBVTOODGHZ-UHFFFAOYSA-N
4-bromobenzaldehyde diethyl acetal,1-bromo-4-diethoxymethyl benzene,4-bromobenzaldehydediethylacetal,benzene, 1-bromo-4-diethoxymethyl,4-bromophenyl diethoxymethane,4-bromobenzaldehyde diethylacetal,4-bromo-alpha,alpha-diethoxytoluene,pubchem5372,acmc-1aigt,4-bromobenzaldehydediethyl acetal
688340
1-bromo-4-(diethoxymethyl)benzene
CCOC(C1=CC=C(C=C1)Br)OCC
Boiling Point 75°C (0.2mmHg)
Specific Gravity 1.274
Density 1.2740g/mL
Flash Point 105°C
Color Colorless to Yellow
Quantity 10g
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5120 to 1.5140 (20°C, 589nm)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula BrC6H4CH(OCH2CH3)2
Formula Weight 259.14
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 4-Bromobenzaldehyde diethyl acetal
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning