Alfa Aesar™ 4-Bromobenzaldehyde dimethyl acetal, 98%

Manufacturer: Alfa Aesar™ H55283AA

Quantity 5mL

 View more versions of this product

Catalog No. AAH55283AA

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24856-58-4
    Formula Weight 231.09
    MDL Number MFCD01320499
    Chemical Name or Material 4-Bromobenzaldehyde dimethyl acetal

    Specifications

    Formula Weight 231.09
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 1.383g/mL
    Boiling Point 254°C
    MDL Number MFCD01320499
    CAS 24856-58-4
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Refractive Index 1.5320
    Quantity 5mL
    Physical Form Liquid
    Chemical Name or Material 4-Bromobenzaldehyde dimethyl acetal
    Assay 98%

    Description

    Applications
    4-Bromobenzaldehyde dimethyl acetal, is used to produce other chemicals, it is used to produce 1-Bromo-4-bromomethyl-benzene at ambient temperature.

    Solubility
    Insoluble in water.

    Notes
    Keep container tightly closed. Store away from oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit