Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Bromobenzamide, 97%

Click to view available options
Quantity:
2g
10g
50g
698-67-9
C7H6BrNO
200.035
MFCD00007991
ZRWNRAJCPNLYAK-UHFFFAOYSA-N
p-bromobenzamide,benzamide, 4-bromo,benzamide, p-bromo,p-bromobenzoic acid amide,4-bromo-benzamide,4-bromo benzamide,pubchem3737,acmc-209obe,4-bromobenzamide,4-bromobenzoic acid amide
69683
4-bromobenzamide
C1=CC(=CC=C1C(=O)N)Br
698-67-9
C7H6BrNO
200.035
MFCD00007991
ZRWNRAJCPNLYAK-UHFFFAOYSA-N
p-bromobenzamide,benzamide, 4-bromo,benzamide, p-bromo,p-bromobenzoic acid amide,4-bromo-benzamide,4-bromo benzamide,pubchem3737,acmc-209obe,4-bromobenzamide,4-bromobenzoic acid amide
69683
4-bromobenzamide
C1=CC(=CC=C1C(=O)N)Br
Melting Point 190°C to 193°C
Quantity 2g
Formula Weight 200.03
Percent Purity 97%
Chemical Name or Material 4-Bromobenzamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning