Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromobenzamide, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
698-67-9
C7H6BrNO
200.03
MFCD00007991
ZRWNRAJCPNLYAK-UHFFFAOYSA-N
p-bromobenzamide, benzamide, 4-bromo, benzamide, p-bromo, p-bromobenzoic acid amide, 4-bromo-benzamide, 4-bromo benzamide, pubchem3737, acmc-209obe, 4-bromobenzamide, 4-bromobenzoic acid amide
69683
4-bromobenzamide
C1=CC(=CC=C1C(=O)N)Br
698-67-9
C7H6BrNO
200.03
MFCD00007991
ZRWNRAJCPNLYAK-UHFFFAOYSA-N
p-bromobenzamide, benzamide, 4-bromo, benzamide, p-bromo, p-bromobenzoic acid amide, 4-bromo-benzamide, 4-bromo benzamide, pubchem3737, acmc-209obe, 4-bromobenzamide, 4-bromobenzoic acid amide
69683
4-bromobenzamide
C1=CC(=CC=C1C(=O)N)Br
Infrared Spectrum Authentic
Color White
Quantity 10g
Packaging Glass bottle
Melting Point 189°C to 194°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula BrC6H4CONH2
Beilstein 09, 353
Formula Weight 200.03
Physical Form Fine Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Bromobenzamide, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-817-9

RTECSNumber : CV2365800