Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromobenzenediazonium tetrafluoroborate, 96%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
673-40-5
C6H4BBrF4N2
270.82
MFCD00011894
FXTCQUCYDJZGGU-UHFFFAOYSA-N
4-bromobenzenediazonium tetrafluoroborate,acmc-209nyn,4-bromobenzene-1-diazonium; tetrafluoroboranuide,boron +3 cation; 4-bromobenzenediazonium; tetrafluoride
2734810
4-bromobenzenediazonium;tetrafluoroborate
[B-](F)(F)(F)F.C1=CC(=CC=C1[N+]#N)Br
673-40-5
C6H4BBrF4N2
270.82
MFCD00011894
FXTCQUCYDJZGGU-UHFFFAOYSA-N
4-bromobenzenediazonium tetrafluoroborate,acmc-209nyn,4-bromobenzene-1-diazonium; tetrafluoroboranuide,boron +3 cation; 4-bromobenzenediazonium; tetrafluoride
2734810
4-bromobenzenediazonium;tetrafluoroborate
[B-](F)(F)(F)F.C1=CC(=CC=C1[N+]#N)Br
Color Beige to Beige-Pink
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Formula Weight 270.82
Physical Form Powder
Percent Purity ≥95%
Chemical Name or Material 4-Bromobenzenediazonium tetrafluoroborate
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger