Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromobenzyl alcohol, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
873-75-6
C7H7BrO
187.04
MFCD00004650
VEDDBHYQWFOITD-UHFFFAOYSA-N
4-bromobenzyl alcohol,4-bromophenyl methanol,p-bromobenzyl alcohol,benzenemethanol, 4-bromo,para-bromobenzyl alcohol,4-bromobenzylalcohol,p-bromo-benzyl alcohol,ccris 5119,4-bromophenyl-methanol,benzyl alcohol, p-bromo
70119
(4-bromophenyl)methanol
C1=CC(=CC=C1CO)Br
873-75-6
C7H7BrO
187.04
MFCD00004650
VEDDBHYQWFOITD-UHFFFAOYSA-N
4-bromobenzyl alcohol,4-bromophenyl methanol,p-bromobenzyl alcohol,benzenemethanol, 4-bromo,para-bromobenzyl alcohol,4-bromobenzylalcohol,p-bromo-benzyl alcohol,ccris 5119,4-bromophenyl-methanol,benzyl alcohol, p-bromo
70119
(4-bromophenyl)methanol
C1=CC(=CC=C1CO)Br
Melting Point 75.0°C to 78.0°C
Color White to Beige
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula BrC6H4CH2OH
Beilstein 06, 446
Formula Weight 187.04
Physical Form Crystals or Crystalline Powder
Percent Purity 99%
Chemical Name or Material 4-Bromobenzyl alcohol, 99%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 212-851-7